Would you like to log in again?
Guinea Pig Complement, MP Biomedicals
Supplier: MP Biomedicals
08642831 08642836
IC642836EA 315.4 USD InStock
IC642836 IC642831
Guinea Pig Complement, MP Biomedicals
Sera (Serum)
The serum is specially collected and processed under "Cold Room" conditions which maintains maximum complement activity
It is suitable for evaluating functional activity of human complement components and for other research requiring a high level of hemolytic activity.
The serum is rapidly collected and frozen-preserving complement activity-and lyophilized
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty