Would you like to log in again?
Freund's Adjuvant, MP Biomedicals
Supplier: MP Biomedicals
08642851 08642861
IC642861EA 298.35 USD InStock
IC642861 IC642851
Freund's Adjuvant, MP Biomedicals
Protein Expression Protein Expression Reagents
Suspended in a mixture of 7.5 mL Arlacel A and 42.5 mL paraffin oil
With or without mycobacterium.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty