Would you like to log in again?
Vector, pUC 18, MP Biomedicals
Supplier: MP Biomedicals
04821226
IC821226EA 255.08 USD InStock
IC821226
Vector, pUC 18, MP Biomedicals
Vectors, Plasmids and Libraries
pUC 18 is a small vector of approximately 2.7 Kb containing the Pvu II/EcoR I fragment of pBR322 and the lacZ gene multiple cloning site from M13mp18.
  • Supplied in 10 mM tris HCl, 1 mM EDTA
  • Clear, colorless solution
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty