Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urea ≥99%, white prills, Ultrapure

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Carbonyl diamideCarbamideCarbonyldiamideDiaminomethanone

04821528 04821519 04821527 04821530
76180-662EA 207.81 USD InStock
76180-662 IC821527 IC821530 IC821519
Urea ≥99%, white prills, Ultrapure
Urea
Urea is the principal end product of nitrogen metabolism in most mammals, formed by the enzymatic reactions of the Kreb's cycle.
Urea is a mild agent usually used in the solubilization and denaturation of proteins. It is also useful for renaturing proteins from samples already denatured with 6 M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex. It is commonly used to solubilize and denature proteins for denaturing isoelectric focusing and two-dimensional electrophoresis and in acetic acid-urea PAGE gels. Urea is used in cell or tissue culture media to increase the osmolality. Urea has also been used as fertilizer because of the easy availability of nitrogen; in animal feeds; it is reacted with aldehydes to make resins and plastics; condensed with malonic ester to form barbituric acid; used in the paper industry to soften cellulose; used as a diuretic; enhances the action of sulfonamides; an antiseptic.
Urea in solution is in equilibrium with ammonium cyanate. The form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins. Carbamylation by isocyanic acid interferes with protein characterization because isocyanic acid reacts with the amino terminus of proteins, preventing N-terminal sequencing. Isocyanic acid also reacts with side chains of lysine and arginine residues resulting in a protein that is unsuitable for many enzymatic digests. In addition, carbamylation often leads to confusing results from peptides having unexpected retention times and masses. When performing enzymatic protein digests it is important to remove urea first. Even though some enzymes will tolerate small amounts of urea, the elevated temperature used for most reactions will lead to carbamylation during the course of the digest. The urea can be removed prior to digestion by fast reversed phase chromatography, spin columns, or dialysis.
Dissolve urea in deionized water to the desired concentration.For every 10 ml of solution, add 1 g of Amberlite® IRA-910.Stir for one hour at room temperature

  • Ultra Pure Grade
  • Partition Coefficient: log Pow: -2,59 - -1,59
  • One gram dissolves in 1 mL water, 10 mL 95% ethanol, 1 mL boiling 95% ethanol, 20 mL absolute ethanol, 6 mL methanol, 2 mL glycerol; Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.
  • Storage temperature: Room Temperature


Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 135 °C (dec.)
Melting Pt: 130…140 °C
Density: 1.335 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 12,10005
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/46/2297046.jpg
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
OD260 (7M aq soln) ≤0.05
OD280 (7M aq soln) ≤0.05
pH (7M aq soln) 8.5 ± 1.0
Conductivity ≤100 uMhos
Copper ≤1ppm
Lead ≤1ppm
Iron ≤1ppm
Cyanate None Detected
RNAse Negative
DNAse Negative
Protease Negative

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty