Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine IgG Isotype Control

Supplier: MP Biomedicals
08641402 08641403
IC8641402EA 399.7 USD InStock
IC8641402 IC8641403
Bovine IgG Isotype Control
Antibodies
This product is useful for structural and biological studies, as a reference standard, or as starting material for immunogen preparation.

  • Prepared using salt fractionation and/or chromatographic techniques
  • Bovine serum
  • Reconstitute with 5.0 ml of 0.15M NaCl for a 2% solution
  • Moisture: 4.30%
  • Purity: 97%
  • ImmunoAssay grade

The immunoglobulin was prepared from bovine serum using current chromatographic techniques. A quantitative protein analysis as well as immunoelectrophoresis and immunodiffusion techniques have been applied to determine quality of the product.
his product may contain a preservative such as sodium azide, thimerosal or proclin. Please see lot specific chemical credential for preservative information.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty