Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
EcoLite(+)™ Liquid Scintillation Cocktail, MP Biomedicals
Supplier: MP Biomedicals
EcoLite(+)™
0188247504 0188247501 0188247505
IC-88247504CS 1727.02 USD InStock
IC-88247504 IC88247501 IC88247505
EcoLite(+)™ Liquid Scintillation Cocktail, MP Biomedicals
Scintillation Cocktails
EcoLite(+)™ is a unique formulation of environmentally safe, nontoxic, and biodegradable media possessing exceptional water and salt holding characteristics.
  • Environmentally Safe
  • Biodegradable
  • Non-Toxic and Non-Flammable
  • Ready-To-Use
  • Versatile
  • Excellent Stability for RIA and Receptor Assays
  • Forms No Gel or Phasing
Difficult-to-count RIA samples, basic samples, phosphate buffer samples, and biological fluids can be counted using EcoLite(+)™, with maximum solubilization resulting in
stable counting solutions. Tritium and C counting efficiency of these samples is exceptionally high.
EcoLite(+)™ is a new generation environmentally safe liquid scintillation cocktail that can be used for virtually all aqueous and non-aqueous samples. It is particularly useful for HPLC fractions, urea, phosphate buffers, protein digests, urine and many other applications where conventional cocktails fail to provide homogeneous counting solutions.
EcoLite(+)™ remains monophasic from 0-30% load capacity. Samples are counted by simply adding them to 10-15 mL of EcoLite(+)™, shaking until clear and counting. NOTE: Temperature and salt concentration will affect sample solubility, as well as, the physical characteristics of a surfactant based cocktail. It is recommended that the counting temperature be considered when determining the proper sample to cocktail ratio. Where phase separation occurs, increase the amount of EcoLite(+)™ or dilute with small amounts of distilled water until the sample clears and remains at the desired counting temperature.
Special Characteristics - EcoLite(+)™ is extremely efficient with both H and C samples. It is very fluid, easily pipetted and mixed without sample up to the maximum load capacity. EcoLite(+)™ does not diffuse into or through polyethylene vials, thereby eliminating erroneous external standard quench measurements due to the vial
wall diffusion effect. It has an extremely low vapor pressure which minimizes volatiles and odors under normal laboratory use. It works well with sodium hydroxide samples up to 0.5 N.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty