Would you like to log in again?
Liver Concentrate Powder from Bovine Liver, MP Biomedicals
Supplier: MP Biomedicals
0290037750 0290037790
IC90037790EA 196.49 USD InStock
IC90037790 IC90037750
Liver Concentrate Powder from Bovine Liver, MP Biomedicals
Specimens Preserved Specimens Preserved Animal Materials
Selectively soluble components of liver extracts were used in early nutritional studies of essential micronutrients.
  • Brown powder
  • Grade NF #XI
  • Foreign activity: E. coli and Salmonella sp. negative
Caution: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. Store at 0°C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty