Would you like to log in again?
HDPE Jugs with White LDPE Foam-Lined Polypropylene Screw Closure, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
315-4000 115-4000 315-2000
IR-315-4000CS 86.12 USD InStock
IR-315-4000 IR115-4000 IR-315-2000
HDPE Jugs with White LDPE Foam-Lined Polypropylene Screw Closure, Thermo Scientific
Bottles Carboys
HDPE jugs meet or exceed EPA performance-based standards for metals, cyanide, and fluoride.
  • Good chemical resistance
  • Good temperature resistance
Jugs are also available in unprocessed form, allowing for customized cleaning procedures.
HDPE jugs feature a narrow mouth and a sturdy handle to make carrying and pouring higher volume samples easy. Recommended for collecting water samples to be analyzed for inorganic trace analytes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty