Would you like to log in again?
I-CHEM™ Cylinder Round Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
013-0250BP 013-1000BP 013-0500BP
IR013-0250BPCS 280.91 USD InStock
IR013-0250BP IR013-1000BP IR-013-0500BP
I-CHEM™ Cylinder Round Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Water Sampling Bottles
Thermo Scientific HDPE Cylinder Rounds are general-purpose containers, supplied bulk and unassembled.
Unprocessed, ready for your own cleaning procedure and assembly with caps purchased separately. These are the same high quality container components as the assembled HDPE cylinder rounds, but sold in bulk, without a cap.
Ordering information: Bulk packed. Closures sold separately, see IR067-2410WBP series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty