Would you like to log in again?
I-CHEM™ Jugs, Amber Glass, with Caps, Thermo Scientific
Supplier: Thermo Scientific
145-4000 245-2360 145-2360 345-4000 345-2360 245-4000
IR245-2360CS 158.25 USD InStock
IR245-2360 IR245-4000 IR345-2360 IR145-2360 IR145-4000 IR345-4000
I-CHEM™ Jugs, Amber Glass, with Caps, Thermo Scientific
Bottles Water Sampling Bottles
For use in the analysis of semivolatiles, pesticides, and PCBs.
  • Great for transporting trip and blank water to the field
  • Excellent for UV light-sensitive liquid samples
I-CHEM Certified™ 200 and 300 series bottles are processed to meet or exceed 'USEPA Specifications and Guidance for Contaminent-Free Containers'. I-CHEM Certified™ 300 series bottles include a Certificate of Analysis with a bar-code production number on every case. Each bottle in the 300 series has a bar-code label for absolute traceability and is compatible with automated sample tracking systems.
I-CHEM™ 100 Series bottles, for general use, are not processed or certified.
Ordering information: Supplied with fluoropolymer resin-lined polypropylene caps attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty