Would you like to log in again?
Open-Top Septa Caps, Thermo Scientific
Supplier: Thermo Scientific
297-2400UB S297-5800 297-2414 S297-3330 S297-5300
IRS297-5800CS 200.32 USD InStock
IRS297-5800 IRS297-3330 IR297-2400UB IR297-2414 IRS297-5300
Open-Top Septa Caps, Thermo Scientific
Closures Bottle Closures
These Open-Top Septa Caps are available with or without a bonded-in PTFE-lined silicone septa
They are the same high-quality polypropylene septa caps used on Thermo Scientific VOA vials, septa bottles, and septa jars. Caps are processed to meet EPA performance-based specifications for Volatile Organic Analysis.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty