Would you like to log in again?
I-CHEM™ High-Density Polyethylene Bottles and Jars, with Caps, Thermo Scientific
Supplier: Thermo Scientific
I-CHEM™
312-0950BPC 312-0500 311-1000 113-0500BPC 319-1000 312-0120BPC 312-0250BPC 312-4000BPC 311-0500 313-0500 113-1000BPC 313-1000 312-0500BPC 312-2000BPC 313-0250 113-0125BPC 319-0125 312-0950 312-4000 312-0250 313-0125 312-2000
IR312-0120BPCCS 1997.43 USD InStock
IR312-0120BPC IR319-0125 IR313-1000 IR313-0500 IR313-0125 IR319-1000 IR312-0250BPC IR312-0500BPC IR312-2000BPC IR312-0950BPC IR312-0500 IR113-0500BPC IR312-4000BPC IR312-0950 IR-312-2000 IR311-0500 IR312-4000 IR312-0250 IR113-1000BPC IR311-1000 IR313-0250 IR113-0125BPC
I-CHEM™ High-Density Polyethylene Bottles and Jars, with Caps, Thermo Scientific
Bottles Water Sampling Bottles
HDPE containers, white, with LDPE-lined polypropylene cap, narrow/wide neck. Suitable for metals analyses and water quality parameters.
  • Narrow mouth for easy handling and precision pouring
  • Wide mouth for easy filling and sample retrieval
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Each container individually barcoded with lot number and unique container number
I-CHEM Certified™ 300 series containers are processed to meet or exceed 'USEPA Specifications and Guidance for Contaminent-Free Containers'. These containers are recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes. Certified to meet EPA performance based specifications for metals, cyanide, fluoride and other water quality parameters. Or unprocessed and readily assembled for own cleaning procedure.
Ordering information: Supplied with polyfoam-lined caps. Also available unprocessed and bulk packed for added economy.
Delivery information: Certificate of analysis (CofA) included in the case and barcoded with lot number for traceability. Analysis labels included unattached for optional use.
Packaging: The Carton is sealed with security tape providing tamper evidence. Each container is individually barcoded with lot number and unique container number.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty