Would you like to log in again?
I-CHEM™ Nalgene® Sample Bottles, High-Density Polyethylene, with Caps, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
N311-1000BPC N311-1000 N311-0500 N311-0125 N319-0125 N311-0250BPC N301-0250 N301-1000 N319-1000 N301-0500 N311-0125BPC N319-0500 N311-0250 N311-0500BPC N301-0125
IRN319-0125CS 358.35 USD InStock
IRN319-0125 IRN319-0500 IRN311-0500BPC IRN311-1000 IRN311-0250 IRN301-0125 IRN311-1000BPC IRN319-1000 IRN301-0500 IRN311-0125 IRN301-1000 IRN311-0500 IRN301-0250 IRN311-0125BPC IRN311-0250BPC
I-CHEM™ Nalgene® Sample Bottles, High-Density Polyethylene, with Caps, Thermo Scientific
Bottles Water Sampling Bottles
Narrow mouth and wide mouth bottles, available in natural and amber, are for use in the analyses of metals.
  • Bottles are precleaned
  • Sample-analysis labels are included, unattached
I-CHEM Certified™ 300 series bottles include a Certificate of Analysis that lists detection limits for specified analyses. Manually readable bar-code labels are attached to each bottle with production number for complete traceability. High-density polyethylene bottles have polypropylene caps attached and are guaranteed leakproof for safe sampling, shipping, and storage.
Ordering information: All containers are supplied with caps attached.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty