Would you like to log in again?
I-CHEM™ Wide Mouth Septa-Jars™, Thermo Scientific
Supplier: Thermo Scientific
S320-0060 S120-0060 S320-0125 S120-0125 S321-0250
IRS320-0125CS 239.96 USD InStock
IRS320-0125 IRS120-0060 IRS320-0060 IRS120-0125 IRS321-0250
I-CHEM™ Wide Mouth Septa-Jars™, Thermo Scientific
Bottles Water Sampling Bottles
Septa-Jars™ are recommended for use in soil headspace analysis and volatile organic analysis.
  • Fluoropolymer resin/silicone septa are bonded into the open-top closures
  • Septa do not reseal after puncture
Short wide mouth jars are excellent for use in Underground Storage Tank (UST) testing.
I-CHEM Certified™ 200 and 300 series jars are processed to meet or exceed U.S. EPA Specifications. I-CHEM Certified™ 300 series jars include a Certificate of Analysis with a bar-coded production number in every case. Each jar in the 300 series has a bar-code label for absolute traceability.
I-CHEM™ 100 Series jars, for general use, are neither processed or certified.
Septa-Jars™ increase the accuracy of analysis by eliminating sample transfer, thereby minimizing volatile loss.
Ordering information: Available in short and tall styles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty