Would you like to log in again?
I-CHEM™ Vials with Screw Cap, Borosilicate Glass, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
S146-0040 C146-0040 TB36-0040 S336-0060 T136-0040 S236-0040 S349-1000 S236-0060 S136-0040 SB46-0040 S126-0020 S146-0060 C136-0040 T336-0040 C246-0040 C126-0020 SB36-0040 S136-0060 T236-0040 S226-0020 T346-0040 S246-0040 S246-0060 S346-0060 S326-0020 S336-0040 S340-0120 C236-0040 C226-0020 T246-0040 C336-0040 C326-0020 S346-0040 C346-0040 C346-0020
IRS236-0060CS 383.49 USD InStock
IRS236-0060 IRS336-0040 IRS236-0040 IRS136-0040 IRS336-0060 IRS136-0060 IRS126-0020 IRT346-0040 IRC136-0040 IRC236-0040 IRC336-0040 IRS246-0040 IRS346-0040 IRS146-0060 IRS246-0060 IRS346-0060 IRSB36-0040 IRS146-0040 IRS340-0120 IRC226-0020 46600-678 IRT246-0040 IRS326-0020 IRC146-0040 IRS226-0020 IRC346-0020 IRC346-0040 IRC246-0040 IRTB36-0040 IRS349-1000 IRC126-0020 IRC326-0020 IRT136-0040 IRT336-0040 IRT236-0040
I-CHEM™ Vials with Screw Cap, Borosilicate Glass, Thermo Scientific
Vials
Thermo Scientific I-Chem Certified™ 200 and 300 series vials meet or exceed U.S
Thermo Scientific I-Chem 100 series vials are made from the same high-quality materials as Thermo Scientific IChem Certified 200 and 300 series, but are not processed and do not include a certificate or container labels. Certificates of Analysis are available by request for 200 series vials and economy packs, but unavailable for the 100 series vials.
EPA 'Specifications and Guidance for Contaminant-Free Sample Containers', and are provided with sample-analysis labels, unattached. Thermo Scientific I-Chem Certified 300 series vials include a Certificate of Analysis with a bar code production number in every case. Each vial in the 300 series has a bar code label for absolute traceability, and is for use with automated or manual sample tracking systems
Ordering information: Available with solid-top fluoropolymer resin-lined screw caps or open-top caps with fluoropolymer resin/silicone septa. The 200 and 300 series vials are packaged in Field-to-Lab tray boxes with partitions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty