Would you like to log in again?
I-CHEM™ Boston Round Bottles, with Open-Top Caps, Thermo Scientific
Supplier: Thermo Scientific
S329-0250 S349-0250 S149-0125 S349-0125 S129-0250
IRS349-0125CS 79.65 USD InStock
IRS349-0125 IRS149-0125 IRS349-0250 IRS329-0250 IRS129-0250
I-CHEM™ Boston Round Bottles, with Open-Top Caps, Thermo Scientific
Bottles Water Sampling Bottles
I-CHEM Certified™ 200 and 300 series glass bottles are processed to meet or exceed 'US EPA Specifications and Guidance for Contaminent-Free Sample Containers' and come with sample-analysis labels, unattached.
I-CHEM™ 100 Series bottles, for general use, are not processed or certified, and do not include sample-analysis labels.
I-CHEM Certified™ 300 series bottles include a Certificate of Analysis with a bar-coded production number in every case. Each bottle in the 300 series has a manually readable bar-code lot label for absolute traceability.
Ordering information: Available in clear or amber glass. Supplied with open-top polypropylene screw caps with 0.125" fluoropolymer resin/silicone septa.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty