Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic anhydride ≥98%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

BAKR0018-01 BAKR0018-03
JT0018-3CS 1712.6 USD InStock
JT0018-3 JT0018-1
Acetic anhydride ≥98%, BAKER ANALYZED® ACS, J.T.Baker®
Acetic anhydride
  • Lot analysis on label.


Formula: (CH₃CO)₂O
MW: 102.09 g/mol
Boiling Pt: 139.9 °C (1013 hPa)
Melting Pt: –73 °C
Density: 1.08 g/cm³ (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8, 3,II
REACH: 01-2119486470-36
Merck Index: 13,00057
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/53/2302253.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃CO)₂O) ≥98 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0020 %
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤3 ppm
Phosphate (PO₄) ≤5 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Heavy Metals (as Pb) ≤2 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Appearance (clear, colorless, volatile liquid)
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty