Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid 99.5-100.5% (dry basis), granular NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

Management of Change (MOC) category = R

BAKR0092-05 BAKR0092-05 BAKR0092-09 BAKR0092-09
JT0092-5CS 2447.68 USD InStock
JT0092-5 JT0092-09 CAJT0092-5 CAJT0092-09
Boric acid 99.5-100.5% (dry basis), granular NF, Multi-Compendial, J.T.Baker®
ortho-Boric acid
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.

Caution: Not for Internal Use


Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now
Specification Test Results
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Solubility in Alcohol Passes Test
NF - Loss on Drying ≤0.5 %
NF - Assay (H₃BO₃) (dried basis) 99.5 - 100.5 %
NF - Endotoxin Concentration (EU/g) ≤35
EP/BP - Assay (H₃BO₃) 99.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 3.8 - 4.8
EP/BP - Solubility in Alcohol Passes Test
EP/BP - Organic Matter Passes Test
EP/BP - Sulfate (SO₄) ≤450 ppm
JP - Assay (H₃BO₃) (dried basis) 99.5 - 101.0 %
JP - Identification Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤5 ppm
JP - Loss on Drying ≤0.5 %
Not for Internal Use
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty