Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citric acid monohydrate 99.0-102.0% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Hydroxytricarballylic acid2-Hydroxy-1,2,3-propanetricarboxilic acid

Management of Change (MOC) category = N

BAKR0110-07 BAKR0110-01 BAKR0110-05 BAKR0110-01 BAKR0110-07 BAKR0110-05
JT0110-7EA 1477.5 USD InStock
JT0110-7 CAJT0110-1 JT0110-5 JT0110-1 CAJT0110-5 CAJT0110-7
Citric acid monohydrate 99.0-102.0% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®
Citric acid monohydrate
The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.


Formula: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Melting Pt: 135…152 °C
Density: 1.54 g/cm³
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://us-legacy.vwr.com/stibo/web/std.lang.all/34/24/713424.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HOC(COOH)(CH₂COOH)₂·H₂O) (by acidimetry) 99.0 - 102.0 %
Insoluble Matter ≤0.005 %
Residue on Ignition ≤0.02 %
Chloride (Cl) ≤0.001 %
Oxalate (C₂O₄) Passes Test
Phosphate (PO₄) ≤0.001 %
Sulfur Compounds (as SO₄) ≤0.002 %
Substances Carbonizable by Hot H₂SO₄ Passes Test
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Lead (Pb) ≤2.000 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty