Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecamolybdophosphoric acid hydrate, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: 12-Molybdophosphoric acid hydrateMolybdophosphoric acid hydratePhosphomolybdic acid hydrate

Management of Change (MOC) category = N

J.T. Baker®
0246-04 BAKR0246-04 0246-01 BAKR0246-01
JT0246-4CS 5920.8 USD InStock
JT0246-4 JT0246-1 CAJT0246-1 CAJT0246-4
Dodecamolybdophosphoric acid hydrate, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Dodecamolybdophosphoric acid hydrate


Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Melting Pt: 78…90 °C
Density: 1.62 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3260
ADR: 8,III
Merck Index: 12,07498
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Insoluble Matter ≤0.01 %
Chloride (Cl) ≤0.02 %
Sulfate (SO₄) ≤0.025 %
Ammonium (NH₄) ≤0.01 %
Calcium (Ca) ≤0.02 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty