Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.25 N, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Muriatic acid

Management of Change (MOC) category = R

BAKR0322-09 BAKR0322-09
JT0322-09EA 7740.01 USD InStock
JT0322-09 CAJT0322-09
Hydrochloric acid 0.25 N, Biotech Reagent, J.T.Baker®
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Ammonium (NH₄) ≤ 3 ppm
Appearance (Clear, colorless liquid) Passes Test
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Normality, Eq/L 0.23 - 0.27
Sulfate (SO₄) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Endotoxin concentration, EU/mL ≤ 1

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty