Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Management of Change (MOC) category = R
BAKR0329-07
BAKR0329-07
BAKR0329-09
BAKR0329-09
BAKR0329-26
JT0329-9EA
6914.29
USD
InStock
JT0329-9
CAJT0329-9
JT0329-26
CAJT0329-7
JT0329-7
Sodium hydroxide 0.5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from USP purified water and sodium hydroxide pellets, NF (Multi-Compendial) which meets BP, Ph. Eur. and JP chemical specifications.