Synonyms:
Muriatic acid
Management of Change (MOC) category = R
J.T. Baker®
BAKR0336-26
BAKR0336-26
0336-03
BAKR0336-03
JT0336-26CS
75445.52
USD
InStock
JT0336-26
CAJT0336-26
JT0336-3
CAJT0336-3
Hydrochloric acid 2.0 N, Biotech Reagent, J.T.Baker®
Hydrochloric acid
Made from USP purified water and hydrochloric acid, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.