Synonyms:
Hydroxytricarballylic acid
Filtered through a 0.2 µm filter.
J.T. Baker®
BAKR0341-07
BAKR0341-07
BAKR0341-25
BAKR0341-27
BAKR0341-26
BAKR0341-26
BAKR0341-27
JT34107EA
1240.12
USD
InStock
JT34107
JT0341-25
JT0341-26
CAJT0341-7
JT0341-27
CAJT0341-26
CAJT0341-27
Citric acid 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Citric acid
Made from purified water USP and citric acid, anhydrous USP/ACS.