Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Sulfosalicylic acid dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: 2-Hydroxy-5-sulphobenzoic acid dihydrate2-Hydroxy-5-sulfobenzoic acid dihydrate

Management of Change (MOC) category = N

BAKR0364-01 BAKR0364-05 BAKR0364-01 BAKR0364-05
JT0364-5CS 7384.48 USD InStock
JT0364-5 CAJT0364-1 CAJT0364-5 JT0364-1
5-Sulfosalicylic acid dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®
5-Sulphosalicylic acid dihydrate
  • Lot analysis on label


Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MW: 254.22 g/mol
Melting Pt: 120 °C
Density: 1.695 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 12,09138
https://us-legacy.vwr.com/stibo/web/std.lang.all/90/04/2299004.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₇H₆O₆S·2H₂O) 99.0 - 101.0 %
Insoluble Matter ≤0.02 %
Residue after Ignition ≤0.10 %
Chloride (Cl) ≤0.001 %
Salicylic Acid (HOC₆H₄COOH) ≤0.04 %
Sulfate (SO₄) ≤0.02 %
Heavy Metals (as Pb) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
STORAGE: Keep product from light.
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty