Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Tartaric acid ≥99.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: L-Tartaric AcidTartaric Acid(2R,3R)-(+)-Tartaric acid(R,R)-(+)-2,3-Dihydroxysuccinic acid(R,R)-(+)-Tartaric acid(2R,3R)-2,3-Dihydroxysuccinic acid

Management of Change (MOC) category = N

BAKR0386-05 BAKR0386-01 BAKR0386-05 BAKR0386-07 BAKR0386-01 BAKR0386-07
JT0386-5CS 3237.59 USD InStock
JT0386-5 CAJT0386-1 JT0386-7 JT0386-1 CAJT0386-5 CAJT0386-7
L(+)-Tartaric acid ≥99.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
L(+)-Tartaric acid
  • Lot analysis on label


Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 168…170 °C
Density: 1.759 g/cm³ (20 °C)
Flash Pt: 150 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 12,09237
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/32/2297232.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HOCO(CHOH)₂COOH) (by acidimetry) ≥99.0 %
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.020 %
Chloride (Cl) ≤0.001 %
Oxalate (C₂O₄) Passes Test
Phosphate (PO₄) ≤0.001 %
Sulfur Compounds (as S) ≤0.002 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty