Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Management of Change (MOC) category = R
BAKR0389-09
BAKR0389-07
BAKR0389-07
BAKR0389-09
JT0389-7EA
1827.19
USD
InStock
CAJT0389-9
CAJT0389-7
JT0389-7
JT0389-9
Sodium hydroxide 1.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from water for injection USP and sodium hydroxide pellets, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.