Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium sulfate octadecahydrate 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Aluminum(III) sulfate octadecahydrateAluminum(III) sulphate octadecahydrate

Management of Change (MOC) category = N

BAKR0564-05 BAKR0564-01 BAKR0564-01 BAKR0564-05
JT0564-5CS 4467.67 USD InStock
CAJT0564-1 JT0564-5 CAJT0564-5 JT0564-1
Aluminium sulfate octadecahydrate 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Aluminium sulphate octadecahydrate
Crystals. Lot analysis on label.


Formula: Al₂(SO₄)₃·18H₂O
MW: 666.43 g/mol
Melting Pt: 86 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00149136
CAS Number: 7784-31-8
EINECS: 233-135-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/26/18570926.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Al₂(SO₄)₃·18H₂O) (by EDTA titrn) 98.0 - 102.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.002 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Potassium (K) ≤0.005 %
ACS - Sodium (Na) ≤0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty