Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycine 98.5-101.5% USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Aminoacetic acid H-Gly-OH

Management of Change (MOC) category = R

BAKR0582-01 BAKR0582-QM BAKR0582-QM BAKR0582-09 0582-05 BAKR0582-09 BAKR0582-01 BAKR0582-07 BAKR0582-07 BAKR0582-05
JT0582-5EA 1177.33 USD InStock
CAJT0582-QM JT0582-5 CAJT0582-1 JT0582-QM JT0582-7 JT0582-9 CAJT0582-5 CAJT0582-7 CAJT0582-9 JT0582-1
Glycine 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
Glycine
Meets USP requirements. Meets BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.


Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg
Order Now
Specification Test Results
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification Passes Test
USP - Loss on Drying at 105°C ≤0.2 %
USP - Residue on Ignition ≤0.1 %
USP - Chloride (Cl) ≤0.007 %
USP - Sulfate (SO₄) ≤0.0065 %
USP - Hydrolyzable Substances Passes Test
USP - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.5 %
USP - Endotoxin Concentration (EU/g) ≤2.5
USP - Related Compounds: Glycine anhydride ≤0.1 %
USP - Related Compounds: Monochloroacetic acid (For Information Only)
USP - Related Compounds: Iminodiacetic acid ≤0.1 %
USP - Related Compounds: Diglycine ≤0.1 %
USP - Related Compounds: Triglycine ≤0.1 %
USP - Related Compounds: Hexamethylenetetramine ≤0.1 %
USP - Related Compounds: Any unspecified impurity (For Information Only)
USP - Related Compounds: Total impurities ≤1.0 %
EP/BP - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 5.9 - 6.4
EP/BP - Chloride (Cl) ≤75 ppm
EP/BP - Loss on Drying at 105°C ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
EP/ BP - Related substances: Glycine anhydride ≤0.10 %
EP/ BP - Related substances: Diglycine ≤0.10 %
EP/ BP - Related substances: Triglycine ≤0.10 %
EP/ BP - Related substances: Unspecified impurities, each ≤0.10 %
EP/ BP - Related substances: Total impurities ≤0.2 %
EP/BP - Ninhydrin Positive Substances - Each Impurity ≤0.10 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤1.0 %
EP/BP - Ammonium (NH₄) ≤0.02 %
JP - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - pH (1 in 20) 5.6 - 6.6
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021 %
JP - Sulfate (SO₄) ≤0.028 %
JP - Ammonium (NH₄) ≤0.02 %
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤0.30 %
JP - Residue on Ignition ≤0.10 %
Only Class 2 Solvents (Methanol) are likely to be present. All are
below Option 1 limits.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty