Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium acetate ≥97%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Acetic acid ammonium salt

Management of Change (MOC) category = NI

J.T. Baker®
BAKR0596-R 0596-07 BAKR0596-07 BAKR0596-01 BAKR0596-01 BAKR0596-R BAKR0596-05 BAKR0596-05
JT0596-REA 8999.91 USD InStock
JT0596-R JT0596-01 JT0596-05 JT0596-7 CAJT0596-R CAJT0596-1 CAJT0596-5 CAJT0596-7
Ammonium acetate ≥97%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium acetate


Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/04/16640604.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃COONH₄) (by acid-base titration) ≥97 %
Insoluble Matter ≤0.005 %
Nitrate (NO₃) ≤0.001 %
pH of 5% Solution at 25°C 6.7 - 7.3
Residue after Ignition ≤0.010 %
Solution (20:100) - clear, colorless, complete Passes Test
Sulfate (SO₄) ≤0.001 %
Water (H₂O) ≤2.5 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty