Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium hydrogen difluoride ≥95.0%, flakes, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Ammonium bifluorideAmmonium hydrogen fluoride

Management of Change (MOC) category = N

BAKR0619-09 BAKR0619-05 BAKR0619-05 BAKR0619-07 BAKR0619-01 BAKR0619-07
JT0619-1CS 479.88 USD InStock
CAJT0619-9 CAJT0619-7 JT0619-1 JT0619-7 JT0619-5 CAJT0619-5
Ammonium hydrogen difluoride ≥95.0%, flakes, BAKER ANALYZED®, J.T.Baker®
Ammonium hydrogen difluoride


Formula: NH₄HF₂
MW: 57.04 g/mol
Melting Pt: 124.6 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (NH₄F·HF) ≥95.0 %
Insoluble Matter ≤0.02 %
pH of 5% Solution at 25°C 4.0 - 6.5
Heavy Metals (as Pb) ≤0.002 %
Moisture (as H₂O) ≤0.4 %
Product may contain black specks that are indigenous to product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty