Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium nitrate ≥95.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

J.T. Baker®
BAKR0729-01 BAKR0729-05 BAKR0729-05 BAKR0729-01 BAKR0729-01 BAKR0729-05
JT0729_1CS 495.76 USD InStock
JT0729_1 JT0729-1 JT0729_5 JT0729-5 CA10781-886 CAJT0729_5
Ammonium nitrate ≥95.0%, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium nitrate


Formula: NH₄NO₃
MW: 80.04 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 169 °C
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011425
CAS Number: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1,III
Merck Index: 12,00567
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NH₄NO₃) ≥95.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - Residue after Ignition ≤0.01 %
ACS - pH of 5% Solution at 25°C 4.5 - 6.0
ACS - Sulfate (SO₄) ≤0.002 %
Chloride (Cl) ≤5 ppm
Nitrite (NO₂) Passes Test
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty