Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium dihydrogen phosphate ≥98.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Ammonium dihydrogenphosphateAmmonium phosphate, monobasicAmmonium Phosphate Monobasic

Management of Change (MOC) category = NI

BAKR0776-09 BAKR0776-R BAKR0776-07 BAKR0776-01 BAKR0776-01 BAKR0776-07 BAKR0776-R
JT0776-7EA 2043.78 USD InStock
JT0776-7 CAJT0776-1 CAJT0776-R CAJT0776-7 CAJT0776-9 JT0776-R JT0776-1
Ammonium dihydrogen phosphate ≥98.0%, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium dihydrogen phosphate


Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Density: 1.80 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NH₄H₂PO₄) ≥98.0 %
Calcium (Ca) ≤0.001 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 3.8 - 4.4
Magnesium (Mg) ≤0.0005 %
Nitrate (NO₃) ≤0.001 %
Sulfate (SO₄) ≤0.01 %
Trace Impurities - Iron (Fe) ≤0.001 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty