Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfate 99.0-100.5% NF endotoxin tested, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Diammonium sulfateSulphuric acid diammonium saltsulfuric acid diammonium saltDiammonium sulphateMascagnite

Management of Change (MOC) category = R

BAKR0798-05 BAKR0798-B1 BAKR0798-05 BAKR0798-SA BAKR0798-B1 BAKR0798-F7 BAKR0798-B2 BAKR0798-F9 BAKR0798-65 BAKR0798-08 BAKR0798-01 BAKR0798-08 BAKR0798-01 BAKR0798-SA BAKR0798-F7 BAKR0798-65 BAKR0798-F9
JT0798-B2EA 8863.26 USD InStock
CAJT0798-B1 JT0798-B2 JT0798-B1 JT0798-F7 CAJT0798-SA JT0798-SA CAJT0798-05 CAJT0798-01 JT0798-05 CAJT0798-65 JT0798-65 JT0798-01 JT0798-8 CAJT0798-8 CAJT0798-F9 JT0798-F9 CAJT0798-F7
Ammonium sulfate 99.0-100.5% NF endotoxin tested, J.T.Baker®
Ammonium sulphate
Preserve in well closed container.


Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.77 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559
Order Now
Specification Test Results
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
NF - Insoluble Matter ≤0.005 %
NF - Residue on Ignition ≤0.005 %
NF - pH of 5% Solution 5.0 - 6.0
NF - Nitrate (NO₃) ≤0.001 %
NF - Chloride (Cl) ≤5 ppm
NF - Phosphate (PO₄) ≤5 ppm
NF - Iron (Fe) ≤5 ppm
NF - Identification Passes Test
NF - Microbial Limit - Total Aerobic Microbial Count (cfu/g) ≤1000
NF - Microbial Limit - Total Combined Molds and Yeasts (cfu/g) ≤10
Endotoxin Concentration (EU/g) ≤2.5
No Class 1, 2 or 3 Residual Solvents are likely to be present by exclusion of the production/purification process.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.
Preserve in well closed container.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty