Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Management of Change (MOC) category = R
BAKR0896-07
BAKR0896-07
JT0896-7EA
2823.98
USD
InStock
JT0896-7
CAJT0896-07
Sodium hydroxide solution 40% (w/w), Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications