Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = R

BAKR0897-QM BAKR0897-07 BAKR0897-09 BAKR0897-QM BAKR0897-79 BAKR0897-07 BAKR0897-09 BAKR0897-79
JT0897-7EA 3011.24 USD InStock
JT0897-7 CAJT0897-79 CAJT0897-07 JT0897-9 CAJT0897-QM CAJT0897-09 JT0897-QM JT0897-79
Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
GMP Manufactured Product
Store above 16°C (60°F) to prevent freezing
CAUTION: For Manufacturing, processing or repackaging
Assay (NaOH) 50 - 52 %
Chloride (Cl) ≤30 ppm
Heavy Metals (as Pb) ≤1 ppm
Iron (Fe) ≤2 ppm
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 5 micron filter.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty