Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium sulfate ≥97.0%, powder, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

BAKR1030-01 BAKR1030-05
JT1030-5CS 7843.29 USD InStock
JT1030-5 JT1030-1
Barium sulfate ≥97.0%, powder, BAKER ANALYZED®, J.T.Baker®
Barium sulphate


Formula: BaSO₄
MW: 233.39 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 1345 °C
Density: 4.50 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (BaSO₄) ≥97.0 %
Acidity (as H₂SO₄) ≤0.01 %
Water-Soluble Salts ≤0.02 %
Chloride (Cl) ≤0.005 %
Phosphate (PO₄) ≤0.001 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - Arsenic (As) ≤1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty