Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bismuth(III) nitrate pentahydrate ≥98.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Bismuth trinitrate pentahydrate

Management of Change (MOC) category = N

BAKR1092-04 BAKR1092-01 BAKR1092-01
JT1092-4CS 835.95 USD InStock
CAJT1092-1 JT1092-4 JT1092-1
Bismuth(III) nitrate pentahydrate ≥98.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Bismuth (III) nitrate pentahydrate


Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 75…80 °C (1013 hPa)
Melting Pt: 30 °C
Density: 2.83 g/cm³
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((Bi(NO₃)₃·5H₂O)) (by EDTA titrn) ≥98.0 %
Insoluble in HNO₃ ≤0.005 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Arsenic (As) ≤0.001 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.002 %
Potassium (K) ≤0.01 %
Silver (Ag) ≤0.001 %
Sodium (Na) ≤0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty