Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride, anhydrous ≥96.0% (by argentometric titration), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Chloro calcium

Management of Change (MOC) category = N

BAKR1311-01 BAKR1311-01 BAKR1311-05 BAKR1311-05 BAKR1311-07 BAKR1311-07
JT1311-5CS 4559.67 USD InStock
CAJT1311-1 JT1311-5 JT1311-7 JT1311-1 CAJT1311-7 CAJT1311-5
Calcium chloride, anhydrous ≥96.0% (by argentometric titration), BAKER ANALYZED® ACS, J.T.Baker®
Calcium chloride
Desiccant, 20 mesh. Lot analysis on label.


Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 13,01660
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CaCl₂) (by Ag titrn) ≥96.0 %
Titrable Base (meq/g) ≤0.006
Mesh - On U.S. No. 20 Sieve ≤10 %
Mesh - Thru U.S. No. 200 Sieve ≤30 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty