Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium oxide, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: LimeQuicklimecausticCalcium oxide from marble

Management of Change (MOC) category = N

BAKR1410-01 BAKR1410-05 BAKR1410-01 BAKR1410-07 BAKR1410-05 BAKR1410-07
JT1410-1CS 412.15 USD InStock
CAJT1410-1 JT1410-1 JT1410-7 JT1410-5 CAJT1410-7 CAJT1410-5
Calcium oxide, BAKER ANALYZED®, J.T.Baker®
Calcium oxide
Powder. Lot analysis on label.


Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C (1013 hPa)
Melting Pt: 2580 °C
Density: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Insoluble in CH₃COOH and NH₄OH Precipitate ≤1.0 %
Loss on Ignition ≤5.0 %
Chloride (Cl) ≤0.005 %
Nitrate (NO₃) ≤0.05 %
Sulfate (SO₄) ≤0.1 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.1 %
Assay, For Information Only

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty