Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium pantothenate 90.0-100.0% USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: D-Calcium pantothenateCalcium D-PantothenateD-Pantothenic acid calcium saltCalcium D-(+)-pantothenateCalcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoateVitamin B5 calcium salt

Management of Change (MOC) category = R

BAKR1443-03 BAKR1443-03 BAKR1443-08 BAKR1443-08
JT1443-8EA 2795.6 USD InStock
JT1443-8 JT1443-3 CAJT1443-8 CAJT1443-3
Calcium pantothenate 90.0-100.0% USP, J.T.Baker®
Calcium pantothenate


Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
Melting Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002766
CAS Number: 137-08-6
EINECS: 205-278-9
Order Now
Specification Test Results
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Identification A Passes Test
Identification B Passes Test
Identification C (Specific Rotation [Å]²⁵ ^D (+)) 25.0 - 27.5 Degree
Alkalinity Passes Test
Loss on Drying ≤5.0 %
Assay (C₁₈H₃₂CaN₂O₁₀)(dried basis) 98.0 - 102.0 %
Content of Calcium (Ca)(dried basis) 8.2 - 8.6 %
Solution Test Passes Test
Only Class 2 solvents (Methanol, Dichloromethane) are likely to be
present. All are below Option 1 limits.
Meets Reagent Specifications for testing USP/NF monographs
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty