Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium sulfate dihydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Plaster of Paris

Management of Change (MOC) category = N

BAKR1452-05 BAKR1452-01 BAKR1452-05
JT1452-5CS 1297.8 USD InStock
JT1452-5 JT1452-1 CAJT1452-5
Calcium sulfate dihydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
Calcium sulphate dihydrate
Powder. Lot analysis on label.


Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CaSO₄·2H₂O) (by EDTA titrn) 98.0 - 102.0 %
Carbonate (CO₃) Passes Test
Chloride (Cl) ≤0.002 %
Heavy Metals (as Pb) ≤0.002 %
Insoluble in Dilute HCl ≤0.02 %
Trace Impurities - Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.02 %
Nitrate (NO₃) Passes Test
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.05 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty