Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromium(III) chloride hexahydrate 99.0-105.0% (by iodometry), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Chromic chloride hexahydrateChromium trichloride hexahydrate

Management of Change (MOC) category = N

1588-05 BAKR1588-R BAKR1588-R BAKR1588-01 BAKR1588-05
JT1588-5CS 7531.79 USD InStock
JT1588-5 JT1588-R JT1588-1 CAJT1588-5 CAJT1588-R
Chromium(III) chloride hexahydrate 99.0-105.0% (by iodometry), BAKER ANALYZED®, J.T.Baker®
Chromium (III) chloride hexahydrate


Formula: CrCl₃·6H₂O
MW: 266.45 g/mol
Melting Pt: 85 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 13,02242
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (CrCl₃·6H₂O) (by iodometry) 99.0 - 105.0 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 2.0 - 3.0
Sulfate (SO₄) ≤0.01 %
Iron (Fe) ≤0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty