Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iron (III) sulfate dodecahydrate 98.5-101.0% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Ferric ammonium sulfate dodecahydrateAmmonium ferric sulfate dodecahydrateAmmonium iron sulfate dodecahydrateFerric ammonium sulphate dodecahydrateAmmonium iron sulphate dodecahydrateAmmonium ferric sulphate dodecahydrate

Management of Change (MOC) category = N

BAKR1988-01 BAKR1988-07 BAKR1988-01 BAKR1988-07 BAKR1988-05 BAKR1988-05
JT1988-7EA 1771.76 USD InStock
JT1988-7 CAJT1988-1 CAJT1988-05 JT1988-1 CAJT1988-7 JT1988-05
Ammonium iron (III) sulfate dodecahydrate 98.5-101.0% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Ammonium iron (III) sulphate dodecahydrate
Crystals. Lot analysis on label.


Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 39…41 °C
Density: 1.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Store in a dry, cool place away from sunlight.
Assay (FeNH₄(SO₄)₂·12H₂O) (by iodometry) 98.5 - 102.0 %
Insoluble Matter ≤0.01 %
Chloride (Cl) ≤0.001 %
Nitrate (NO₃) ≤0.01 %
Calcium (Ca) ≤0.01 %
Copper (Cu) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Zinc (Zn)(by AAS) ≤0.003 %
Ferrous Iron Passes Test
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty