Synonyms:
Ferric ammonium sulfate dodecahydrate
, Ammonium ferric sulfate dodecahydrate
, Ammonium iron sulfate dodecahydrate
, Ferric ammonium sulphate dodecahydrate
, Ammonium iron sulphate dodecahydrate
, Ammonium ferric sulphate dodecahydrate
Management of Change (MOC) category = N
BAKR1988-01
BAKR1988-07
BAKR1988-01
BAKR1988-07
BAKR1988-05
BAKR1988-05
JT1988-7EA
1771.76
USD
InStock
JT1988-7
CAJT1988-1
CAJT1988-05
JT1988-1
CAJT1988-7
JT1988-05
Ammonium iron (III) sulfate dodecahydrate 98.5-101.0% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Ammonium iron (III) sulphate dodecahydrate
Crystals. Lot analysis on label.