Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Asparagine monohydrate NF, Multi-Compendial, J.T.Baker®

Supplier: Avantor Performance Materials
Synonyms: H-Asn-OH.H2OH-Asn-OH·H₂OL-Asparagine monohydrate(S)-(+)-Asparagine monohydrate(S)-2,4-Diamino-4-oxobutanoic acid hydrate

BAKR2055-09 BAKR2055-07 BAKR2055-06
JT2055-09EA 23572.51 USD InStock
JT2055-09 JT2055-07 JT2055-06
L(+)-Asparagine monohydrate NF, Multi-Compendial, J.T.Baker®
L(+)-Asparagine monohydrate
US sourced amino acids

Caution: For Manufacturing, processing or repackaging


Formula: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 215…217 °C
Density: 1.567 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9
Merck Index: 13,00842
Order Now
Specification Test Results
NF - Assay (dried basis) 95.5 - 102.0 %
NF - Identification A Passes test
NF - Identification B Passes test
NF - Residue on Ignition ≤ 0.1 %
NF - Lead (Pb) ≤ 5 ppm
NF - Chromatographic Purity: Asparagine Related Compound A ≤ 1.0 %
NF - Chromatographic Purity: Aspartic acid ≤ 1.0 %
NF - Chromatographic Purity: Each individual unidentified impurity ≤ 0.5 %
NF - Chromatographic Purity: Total Impurities ≤ 3.0 %
NF - Specific Rotation @ 20°C (+) 33.0 - 36.5 °
NF - Total Aerobic Microbial Count (cfu/g) ≤ 1000
NF - Total Yeast and Mold Count (cfu/g) ≤ 100
NF - Loss on Drying at 130°C 11.5 - 12.5 %
EP/BP - Assay (dried basis) 99.0 - 101.0 %
EP/BP - Identification A Passes test
EP/BP - Identification B Passes test
EP/BP - Identification D Passes test
EP/BP - Appearance of Solution Passes test
EP/BP - pH 4.0 - 6.0
EP/BP - Specific Rotation @ 20°C (+) 33.7 - 36.0 °
EP/BP - Related Substances - Impurity A ≤ 0.5 %
EP/BP - Related Substances - Impurity C ≤ 0.1 %
EP/BP - Related Substances - Unspecified Impurities, each ≤ 0.05 %
EP/BP - Related Substances - Total impurities ≤ 0.8 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 200 ppm
EP/BP - Ammonium (NH₄) ≤ 0.1 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on drying (130°C) 10.5 - 12.5 %
EP/BP - Sulphated ash ≤ 0.1 %
Endotoxin ≤ 10 EU/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty