Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Arginine hydrochloride USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: (S)-(+)-Arginine hydrochlorideL-ArgL-Arginine, hydrochloride(S)-2-Amino-5-guanidinopentanoic acid hydrochlorideL(+)-Arginine hydrochlorideL(+)-Arginine monohydrochloride(S)-(+)-Arginine HClH-L-Arg-OH.HClH-Arg-OH.HCl

BAKR2067-06 BAKR2067-QM BAKR2067-06 BAKR2067-07 BAKR2067-09 BAKR2067-QM BAKR2067-07 BAKR2067-09
JT2067-6EA 532.16 USD InStock
JT2067-6 JT2067-7 JT2067-9 CAJT2067-9 CAJT2067-7 CAJT2067-6 CAJT2067-QM JT2067-QM
L-Arginine hydrochloride USP, Multi-Compendial, J.T.Baker®
L-Arginine hydrochloride


Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 218 °C
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Order Now
Specification Test Results
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
Preserve in well-closed containers.
Store in well-closed,light-resistant containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²º^D (+) 21.4 - 23.6 Degree
USP - Loss on Drying at 105°C ≤0.2 %
USP - Residue on Ignition ≤0.1 %
USP - Sulfate (SO₄) ≤0.03 %
USP - Related Compounds - Individual Impurities ≤0.5 %
USP - Related Compounds - Total Impurities ≤2.0 %
USP - Chloride (Cl) 16.5 - 17.1 %
EP/BP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 21.0 - 23.5 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5 %
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤0.02 %
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation (+) 21.5 - 23.5 Degree
JP - pH (1 in 10) 4.7 - 6.2
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) ≤0.028 %
JP - Ammonium (NH₄) ≤0.02 %
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤0.20 %
JP - Residue after Ignition ≤0.10 %
Endotoxin Concentration, IU/mg, For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty