Synonyms:
Ferrous sulphate heptahydrate
, Iron vitriol
, Ferrous sulfate heptahydrate
Management of Change (MOC) category = N
BAKR2070-07
BAKR2070-05
BAKR2070-01
BAKR2070-05
BAKR2070-R
BAKR2070-R
BAKR2070-01
BAKR2070-07
JT2070-REA
8146.07
USD
InStock
CAJT2070-7
JT2070-R
JT2070-1
CAJT2070-1
JT2070-7
JT2070-5
CAJT2070-5
CAJT2070-R
Iron(II) sulfate heptahydrate 99.0-104.5% (by KMnO₄ titration), BAKER ANALYZED® ACS, J.T.Baker®
Iron (II) sulphate heptahydrate
Granular. Lot analysis on label.
Caution: Fluctuating temperatures cause product oxidation.
IMPORTANT: Store below 24°C (75°F).