Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Methionine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acidL(-)-MethionineMethionine(S)-(-)-2-Amino-4-(methylthio)butanoic acidH-Met-OH(S)-(-)-Methionine(S)-2-Amino-4-(methylthio)butanoic acid

Management of Change (MOC) category = R

J.T. Baker®
BAKR2085-05 BAKR2085-07 BAKR2085-06 BAKR2085-07 BAKR2085-06 BAKR2085-05
JT2085-07CS 10032.24 USD InStock
CAJT2085-6 CAJT2085-5 JT2085-07 CAJT2085-07 JT2085-6 JT2085-5
L-Methionine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
L-Methionine
  • Food GMP manufactured product


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 136 °C
Density: 1.373 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004
Order Now
Specification Test Results
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₅H₁₁NO₂S) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 22.4 - 24.7 Degree
USP - pH 5.6 - 6.1
USP - Loss on Drying at 105°C ≤0.3 %
USP - Residue on Ignition ≤0.4 %
USP - Chloride (Cl) ≤0.05 %
USP - Sulfate (SO₄) ≤0.03 %
USP - Iron (Fe) ≤0.003 %
USP - Related Compounds - Methionine sulfoxide ≤0.1 %
USP - Related Compounds - N-Acetyl-D,L- methionine ≤0.2 %
USP - Related Compounds - Any unspecified impurity ≤0.1 %
USP - Related Compounds - Total unspecified impurities ≤0.30 %
EP/BP - Assay (C₅H₁₁NO₂S) (dried basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 5.5 - 6.5
EP/BP - Specific Rotation [Å]²º^D (+) 22.5 - 24.0 Degree
EP/BP - Related Substances: Unspecified Impurities, each ≤0.10 %
EP/BP - Related Substances: Total Impurities ≤0.3 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤200 ppm
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying at 105°C ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (C₅H₁₁NO₂S) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation (+) 21.0 - 25.0 Degree
JP - pH 5.2 - 6.2
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021 %
JP - Sulfate (SO₄) ≤0.028 %
JP - Ammonium (NH₄) ≤0.02 %
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤0.30 %
JP - Residue after Ignition ≤0.10 %
Endotoxin Concentration, IU/mg, For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Only Class 2 Solvents (Methanol) are likely to be present. All are
below Option 1 limits.
Elemental Impurities (USP (232), EP 5.20) - Information on
elemental impurities for this product is available on the
associated Product Regulatory Data Sheet and elemental impurity
profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty