Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Threonine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: (2S,3R)-(-)-Threonine(2S,3R)-2-Amino-3-hydroxybutyric acidH-Thr-OH

Management of Change (MOC) category = R

BAKR2089-06 BAKR2089-06 BAKR2089-07 BAKR2089-07
JT2089-7EA 26990.66 USD InStock
JT2089-7 JT2089-6 CAJT2089-6 CAJT2089-7
L-Threonine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
L-Threonine
  • Food GMP manufactured product


Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1
Order Now
Specification Test Results
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Store in well-closed,light-resistant containers
Preserve in well-closed containers.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₄H₈NO₃) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D -29.1 - -26.7 Degree
USP - pH 5.0 - 6.5
USP - Loss on Drying at 105°C ≤0.2 %
USP - Residue on Ignition ≤0.4 %
USP - Chloride (Cl) ≤0.05 %
USP - Sulfate (SO₄) ≤0.03 %
USP - Iron (Fe) ≤0.003 %
USP - Related Compounds - Individual Impurities ≤0.5 %
USP - Related Compounds - Total Impurities ≤2.0 %
EP/BP - Assay (C₄H₈NO₃) (dried basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 5.0 - 6.5
EP/BP - Specific Rotation [Å]²º^D -29.0 - -27.6 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤0.02 %
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (C₄H₈NO₃) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation -29.0 - -26.0 Degree
JP - pH 5.2 - 6.2
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021 %
JP - Sulfate (SO₄) ≤0.028 %
JP - Ammonium (NH₄) ≤0.02 %
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤0.20 %
JP - Residue on Ignition ≤0.10 %
Endotoxin Concentration, IU/mg, For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty